Learn More
Enzo Life Sciences H-Arg-AMC. 2HCl (25mg)
SDP
Catalog No. 502011245
Encompass_Preferred
Fluorogenic substrate for Cathepsin H (Km=150 µM) and aminopeptidases. Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144). Purity: ≥99%. Long Term Storage: -20°C.
Catalog No. 50-201-1245 Supplier Enzo Life Sciences Supplier No. BMLP1350025
Fluorogenic substrate for Cathepsin H (Km=150 µM) and aminopeptidases. Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144). Purity: ≥99%. Long Term Storage: -20°C.