Learn More
Bachem H-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-OH Trifluoroacetate
SDP
Supplier:  Bachem 4040952.0005
Encompass_Preferred
H-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-OH Trifluoroacetate, 143413-47-2, C54H110N36O10, Mr 1423.70, 5mg. H(-Arg)9-OH, Nonaarginine, (Arg)9. Nonaarginine is one of the most efficient protein transduction domains and a cell-penetrating peptide.
Catalog No. 50-260-056