Learn More
Bachem H-Arg-Gly-Asp-Ser-OH
SDP
Supplier:  Bachem 4008997.0025
Encompass_Preferred
H-Arg-Gly-Asp-Ser-OH, 91037-65-9, C15H27N7O8, Mr 433.42, 25mg. RGDS. RGDS, a tetrapeptide which supports fibroblast attachment and inhibits fibronectin binding to platelets. RGDS is a target sequence for syphilis spirochete cytadherence.
Catalog No. 50-259-468