Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Arg-Lys-OH TFA | 99.86% | 10 MG
SDP

Catalog No. 5000339322
Encompass_Preferred

H-Arg-Lys-OH TFA is a dipeptide formed from L-arginyl and L-lysine residues. It is an endogenous metabolite and is for research use only. This product is involved in the characterization of physiologically relevant advanced glycation end products (AGEs), including lysine-lysine and lysine-arginine cross-links, which have been characterized from tissues ex vivo.

  • Dipeptide formed from L-arginyl and L-lysine residues
  • Endogenous metabolite
  • For research use only
  • Involved in advanced glycation end products (AGEs) characterization
  • Soluble in H2O
  • Molecular weight of 416.40
  • Formula: C14H27F3N6O5
  • Appearance: Solid, white to off-white
  • Purity: 99.86%

Catalog No. 50-003-39322 Supplier Medchemexpress LLC Supplier No. HY126487A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Arg-Lys-OH TFA is a dipeptide formed from L-arginyl and L-lysine residues. It is an endogenous metabolite and is for research use only. This product is involved in the characterization of physiologically relevant advanced glycation end products (AGEs), including lysine-lysine and lysine-arginine cross-links, which have been characterized from tissues ex vivo.

  • Dipeptide formed from L-arginyl and L-lysine residues
  • Endogenous metabolite
  • For research use only
  • Involved in advanced glycation end products (AGEs) characterization
  • Soluble in H2O
  • Molecular weight of 416.40
  • Formula: C14H27F3N6O5
  • Appearance: Solid, white to off-white
  • Purity: 99.86%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.