Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Arg-Lys-OH (TFA) | 99.86% | 25 MG
SDP

Catalog No. 5000342812
Encompass_Preferred

H-Arg-Lys-OH TFA is a dipeptide composed of L-arginyl and L-lysine residues. It is classified as an endogenous metabolite. Research indicates that advanced glycation end products (AGEs), particularly those forming lysine-lysine and lysine-arginine cross-links, are physiologically relevant and have been characterized from ex vivo tissues.

  • Dipeptide formed from L-arginyl and L-lysine residues.
  • Endogenous metabolite.
  • Used for research purposes only.
  • Available in solid and solution forms.
  • Solubility in H2O: 125 mg/mL (300.19 mM; requires sonication).
  • Storage conditions: -20°C, protected from light, under nitrogen (solid); -80°C for 6 months or -20°C for 1 month (in solvent, protected from light, under nitrogen).

Catalog No. 50-003-42812 Supplier Medchemexpress LLC Supplier No. 25MGHY126487A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Arg-Lys-OH TFA is a dipeptide composed of L-arginyl and L-lysine residues. It is classified as an endogenous metabolite. Research indicates that advanced glycation end products (AGEs), particularly those forming lysine-lysine and lysine-arginine cross-links, are physiologically relevant and have been characterized from ex vivo tissues.

  • Dipeptide formed from L-arginyl and L-lysine residues.
  • Endogenous metabolite.
  • Used for research purposes only.
  • Available in solid and solution forms.
  • Solubility in H2O: 125 mg/mL (300.19 mM; requires sonication).
  • Storage conditions: -20°C, protected from light, under nitrogen (solid); -80°C for 6 months or -20°C for 1 month (in solvent, protected from light, under nitrogen).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.