Learn More
Bachem H-Arg-Phe-OH
SDP
Supplier:  Bachem 4002139.1000
Encompass_Preferred
H-Arg-Phe-OH Acetate, 2047-13-4, C15H23N5O3, Mr 321.38, 1g. The dipeptide Arg-Phe mimics the haemodynamic effects of FMRF amide. Both peptides increased blood pressure and heart rate in anaesthesized rats by central stimulation of the sympathetic nervous system, RF being approximately 4 fold more potent.
Catalog No. 50-382-014