Learn More
Bachem H-Arg-Pro-OH
SDP
Supplier:  Bachem 4011377.0250
Encompass_Preferred
H-Arg-Pro-OH Sulfate, 2418-69-1, C11H21N5O3, Mr 271.32, 250mg. Dipeptide corresponding to the N-terminal sequence of substance P. During SP metabolism, H-Arg-Pro-OH and H-Lys-Pro-OH are split off by post-proline dipeptidyl aminopeptidase (PPDAP), an enzyme expressed in the brain.
Catalog No. 50-382-295