Learn More
CPC Scientific H-Asn-Pro-Glu-Tyr(PO3H2)-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ILGF001A
Encompass
The phosphorylated tetrapeptide NPEpY represents a substrate binding domain common to the insulin-like growth factor-I receptor (IGF-IR) and the insulin receptor (IR). Substrate binding to this motif depends on autophosphorylation by the receptor tyrosine kinaseONE-LETTER SEQUENCE: NPEr(PO3H2)YMOLECULAR FORMULA: C23H32N5O12P1MOLECULAR WEIGHT:601.51STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [290810-63-8], RESEARCH AREA: Diabetes
Catalog No. 50-210-9583