Learn More
Medchemexpress LLC H-Asp(AMC)-OH | 133628-73-6 | MFCD00236804 | 99.8% | 290.27 g/mol | C14H14N2O5 | 10 MG
SDP
Catalog No. 5000283693
Encompass_Preferred
H-Asp(AMC)-OH is an aspartic acid derivative linked to 7-amino-4-methylcoumarin (AMC) used as a fluorescent substrate in biochemical and enzymatic assays. It is supplied in small research quantities and characterized for assay use with reported high purity and defined storage recommendations.
  • Fluorescent aspartic acid derivative for use as a substrate in assays.
  • Does not inhibit glycine transport at a concentration of 0.25 mM.
  • High purity (99.75%) suitable for research applications.
  • Molecular weight 290.27 g/mol; formula C14H14N2O5.
  • Available in a 10 mg pack size for laboratory use.
  • Store sealed, away from moisture and light; in solvent: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-002-83693 Supplier Medchemexpress LLC Supplier No. HYW14211710MG
H-Asp(AMC)-OH is an aspartic acid derivative linked to 7-amino-4-methylcoumarin (AMC) used as a fluorescent substrate in biochemical and enzymatic assays. It is supplied in small research quantities and characterized for assay use with reported high purity and defined storage recommendations.
  • Fluorescent aspartic acid derivative for use as a substrate in assays.
  • Does not inhibit glycine transport at a concentration of 0.25 mM.
  • High purity (99.75%) suitable for research applications.
  • Molecular weight 290.27 g/mol; formula C14H14N2O5.
  • Available in a 10 mg pack size for laboratory use.
  • Store sealed, away from moisture and light; in solvent: -80°C for 6 months, -20°C for 1 month.