Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Asp(AMC)-OH | 133628-73-6 | MFCD00236804 | 99.8% | 290.27 g/mol | C14H14N2O5 | 5 MG
SDP

Catalog No. 5000283782
Encompass_Preferred

H-Asp(AMC)-OH is an amino acid derivative bearing a 7-amino-4-methylcoumarin (AMC) fluorophore, provided as a solid for use as a fluorescent substrate or dye in biochemical assays. It has defined molecular properties and is supplied with high reported purity suitable for research applications.

  • Fluorescent AMC-labeled aspartic acid derivative.
  • High purity (≈99.8%).
  • Molecular weight 290.27 g/mol.
  • Molecular formula C14H14N2O5.
  • Solid, white to off-white powder.
  • Available in milligram packaging for assay use.
  • Reported not to inhibit glycine transport at 0.25 mM.

Catalog No. 50-002-83782 Supplier Medchemexpress LLC Supplier No. HYW1421175MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Asp(AMC)-OH is an amino acid derivative bearing a 7-amino-4-methylcoumarin (AMC) fluorophore, provided as a solid for use as a fluorescent substrate or dye in biochemical assays. It has defined molecular properties and is supplied with high reported purity suitable for research applications.

  • Fluorescent AMC-labeled aspartic acid derivative.
  • High purity (≈99.8%).
  • Molecular weight 290.27 g/mol.
  • Molecular formula C14H14N2O5.
  • Solid, white to off-white powder.
  • Available in milligram packaging for assay use.
  • Reported not to inhibit glycine transport at 0.25 mM.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.