Learn More
Medchemexpress LLC H-Asp(Obzl)-OtBu HCl | 52615-97-1 | MFCD00190761 | 100.0% | 315.79 g/mol | C15H22ClNO4 | 10 G
SDP
Catalog No. 5000209129
Encompass_Preferred
H-Asp(Obzl)-OtBu HCl is a protected aspartic acid derivative supplied as the hydrochloride salt. It serves as a building block for peptide and organic synthesis, providing side-chain benzyl (Obzl) protection and a tert-butyl ester on the carboxyl group to enable orthogonal deprotection strategies.
  • Protected amino acid for peptide synthesis and organic synthesis
  • Side-chain benzyl (Obzl) protection reduces side reactions during coupling
  • Tert-butyl ester enables orthogonal deprotection strategies
  • Supplied as the hydrochloride salt for handling convenience and stability
  • Solid, white to off-white, high-purity material suitable for synthesis
Catalog No. 50-002-09129 Supplier Medchemexpress LLC Supplier No. HYW04199610G
H-Asp(Obzl)-OtBu HCl is a protected aspartic acid derivative supplied as the hydrochloride salt. It serves as a building block for peptide and organic synthesis, providing side-chain benzyl (Obzl) protection and a tert-butyl ester on the carboxyl group to enable orthogonal deprotection strategies.
  • Protected amino acid for peptide synthesis and organic synthesis
  • Side-chain benzyl (Obzl) protection reduces side reactions during coupling
  • Tert-butyl ester enables orthogonal deprotection strategies
  • Supplied as the hydrochloride salt for handling convenience and stability
  • Solid, white to off-white, high-purity material suitable for synthesis