Learn More
Medchemexpress LLC H-Asp(Obzl)-OtBu HCl | 52615-97-1 | MFCD00190761 | 100.0% | 315.79 g/mol | C15H22ClNO4 | 100 G
SDP
Catalog No. 5000208822
Encompass_Preferred
H-Asp(Obzl)-OtBu·HCl is a protected aspartic acid derivative supplied as the hydrochloride salt and used as an amino acid building block in peptide synthesis and synthetic organic chemistry.
  • Protected side-chain with benzyl (Obzl) group
  • Tert-butyl ester (OtBu) on the side chain
  • Provided as the hydrochloride salt for stability and handling
  • High purity suitable for research applications
  • Available in multiple package sizes, including 10 G, 25 G, 100 G
  • Molecular weight 315.79 g/mol, formula C15H22ClNO4
Catalog No. 50-002-08822 Supplier Medchemexpress LLC Supplier No. HYW041996100G
H-Asp(Obzl)-OtBu·HCl is a protected aspartic acid derivative supplied as the hydrochloride salt and used as an amino acid building block in peptide synthesis and synthetic organic chemistry.
  • Protected side-chain with benzyl (Obzl) group
  • Tert-butyl ester (OtBu) on the side chain
  • Provided as the hydrochloride salt for stability and handling
  • High purity suitable for research applications
  • Available in multiple package sizes, including 10 G, 25 G, 100 G
  • Molecular weight 315.79 g/mol, formula C15H22ClNO4