Learn More
Perkin Elmer US LLC Torch Module Mounting Block for Optima 2x00/4x00/5x00/7x00 DV/8x00
Catalog No. 509043794
Encompass_Preferred
This torch module mounting block is a sample introduction component used with Optima™ 2x00 DV, 4x00 DV, 5x00 DV, 7x00 DV and 8x00 ICP-OES series instruments. It is used in the Quick-Change Torch Module with Cyclonic Spray Chamber and Concentric Nebulizer for Optima 2x00/4x00/5x00/7x00(N0770607) and for Optima 8x00 (N0780607), and the Quick-Change Torch Module with Scott Spray Chamber and GemTip Cross-Flow II Nebulizer for Optima 2x00/4x00/5x00/7x00 (N0770606).
Catalog No. 50-904-3794 Supplier Perkin Elmer US LLC Supplier No. N0770600
This torch module mounting block is a sample introduction component used with Optima™ 2x00 DV, 4x00 DV, 5x00 DV, 7x00 DV and 8x00 ICP-OES series instruments. It is used in the Quick-Change Torch Module with Cyclonic Spray Chamber and Concentric Nebulizer for Optima 2x00/4x00/5x00/7x00(N0770607) and for Optima 8x00 (N0780607), and the Quick-Change Torch Module with Scott Spray Chamber and GemTip Cross-Flow II Nebulizer for Optima 2x00/4x00/5x00/7x00 (N0770606).