Learn More
Apexbio Technology LLC H-Cit-OH 372-75-8 100g
SDP
Catalog No. 5000097284
Encompass_Preferred
H-Cit-OH (CAS 372-75-8) also known as L-citrulline is a non-proteinogenic amino acid produced as an intermediate in the urea cycle Within cellular metabolism L-citrulline participates in nitrogen homeostasis by facilitating ammonia detoxification and is further converted to L-arginine via the ornithine cycle thereby promoting nitric oxide (NO) synthesis It is widely utilized in biomedical research to investigate amino acid metabolic pathways regulation of nitrogen balance and modulation of cardiovascular physiology Additionally L-citrulline serves as a metabolic precursor in cell culture models to study signaling pathways and related biological processes
Catalog No. 50-000-97284 Supplier Apexbio Technology LLC Supplier No. A7486100000
H-Cit-OH (CAS 372-75-8) also known as L-citrulline is a non-proteinogenic amino acid produced as an intermediate in the urea cycle Within cellular metabolism L-citrulline participates in nitrogen homeostasis by facilitating ammonia detoxification and is further converted to L-arginine via the ornithine cycle thereby promoting nitric oxide (NO) synthesis It is widely utilized in biomedical research to investigate amino acid metabolic pathways regulation of nitrogen balance and modulation of cardiovascular physiology Additionally L-citrulline serves as a metabolic precursor in cell culture models to study signaling pathways and related biological processes