Learn More
Prolume Ltd h-Coelenterazine-SOL (in vivo), 1 mg
Catalog No. NC1937868
Encompass
Water-soluble h-Coelenterazine is formulated to be water-soluble and safe for in vivo imaging applications with Renilla Luciferase.
Catalog No. NC1937868 Supplier Prolume Ltd Supplier No. 30111
Water-soluble h-Coelenterazine is formulated to be water-soluble and safe for in vivo imaging applications with Renilla Luciferase.