Learn More
Bachem H-Cys(aminoethyl)-OH · HCl
SDP
Supplier:  Bachem 4001430.0001
Encompass_Preferred
H-Cys(aminoethyl)-OH · HCl, 4099-35-8, C5H12N2O2S · HCl, Mr 200.69, 1g. L-4-Thialysine · HCl, L-Thiosine · HCl. A naturally occuring cytotoxic lysine analog. Substrate of bacterial lysine-2-monooxygenase. L-Thialysine inhibits homocitrate synthase, E. coli aspartate kinase III, and the lysine aminomutases lysine-2,3-aminomutase and lysine-5,6-aminomutase.
Catalog No. 50-397-445