Learn More
Bachem H-Cys(NPys)-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-NH2
SDP
Catalog No. NC1700221
Encompass_Preferred
H-Cys(NPys)-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-NH2 Trifluoroacetate, 1676104-79-2, C62H118N40O12S2, Mr 1680.01, 1mg. Cys(Npys)-(D-Arg)9. All D-analog of cell-permeable peptide C(Npys)RRRRRRRRR-NH2.
Catalog No. NC1700221 Supplier Bachem Supplier No. 4091590.0001
H-Cys(NPys)-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-D-Arg-NH2 Trifluoroacetate, 1676104-79-2, C62H118N40O12S2, Mr 1680.01, 1mg. Cys(Npys)-(D-Arg)9. All D-analog of cell-permeable peptide C(Npys)RRRRRRRRR-NH2.