Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-Cys Trt -OH, Advanced ChemTech

Catalog No. 01339689
Click to view available options
Quantity:
1 g
5 g
25 g

Catalog No. 01-339-689

Supplier: Advanced Chem Tech IC720625G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2799-07-7
Molecular Formula C22H21NO2S
Molecular Weight (g/mol) 363.48
InChI Key DLMYFMLKORXJPO-UHFFFAOYNA-N
IUPAC Name 2-amino-3-[(triphenylmethyl)sulfanyl]propanoic acid
SMILES NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Specifications

Color White/Off-White
Quantity 25 g
Grade Biochemical
Chemical Name or Material H-Cys Trt -OH
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.