Learn More
Medchemexpress LLC H-D-2-Nal-OH | 76985-09-6 | 215.25 | 10 G
SDP
Catalog No. 5000447407
Encompass_Preferred
H-D-2-Nal-OH is an alanine derivative that influences the secretion of anabolic hormones, supplies fuel during exercise, improves mental performance during stress-related tasks, and helps prevent exercise-induced muscle damage. It is recognized as a beneficial ergogenic dietary substance.
  • Solid appearance
  • White to off-white color
  • Powder stable for 3 years at -20°C, 2 years at 4°C
  • Soluble in water (1 mg/mL)
Catalog No. 50-004-47407 Supplier Medchemexpress LLC Supplier No. HYW00826910G
H-D-2-Nal-OH is an alanine derivative that influences the secretion of anabolic hormones, supplies fuel during exercise, improves mental performance during stress-related tasks, and helps prevent exercise-induced muscle damage. It is recognized as a beneficial ergogenic dietary substance.
  • Solid appearance
  • White to off-white color
  • Powder stable for 3 years at -20°C, 2 years at 4°C
  • Soluble in water (1 mg/mL)