Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-D-Ala-OMe hydrochloride | 14316-06-4 | 139.58 | 500 G
SDP

Catalog No. 5000446390
Encompass_Preferred

H-D-Ala-OMe hydrochloride is an alanine derivative intended for research use. This amino acid derivative is commercially used as an ergogenic supplement, influencing anabolic hormone secretion, fuel supply during exercise, mental performance under stress, and helping prevent exercise-induced muscle damage.

  • Purity: 98.0%
  • Appearance: Solid
  • Color: White to off-white
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
  • Solubility (in vitro): DMSO: 100 mg/mL (716.44 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)

Catalog No. 50-004-46390 Supplier Medchemexpress LLC Supplier No. HYW007986500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-D-Ala-OMe hydrochloride is an alanine derivative intended for research use. This amino acid derivative is commercially used as an ergogenic supplement, influencing anabolic hormone secretion, fuel supply during exercise, mental performance under stress, and helping prevent exercise-induced muscle damage.

  • Purity: 98.0%
  • Appearance: Solid
  • Color: White to off-white
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
  • Solubility (in vitro): DMSO: 100 mg/mL (716.44 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.