Learn More
Medchemexpress LLC H-D-Asp(OtBu)-OH | 64960-75-4 | MFCD00077101 | 99.9% | 189.21 | C8H15NO4 | 5 G
SDP
Catalog No. 5000302561
Encompass_Preferred
H-D-Asp(OtBu)-OH is the tert-butyl ester-protected D-aspartic acid with a free N-terminus, supplied as a high-purity solid for use as a peptide synthesis building block and in organic synthesis.
  • Protected D-aspartic acid for solid-phase and solution-phase peptide synthesis.
  • High purity suitable for research and development applications.
  • Tert-butyl side-chain protection provides stability during coupling and deprotection workflows.
  • Available in small lab pack sizes to support method development and scale-up.
  • Well characterized: CAS 64960-75-4; molecular weight 189.21; formula C8H15NO4.
Catalog No. 50-003-02561 Supplier Medchemexpress LLC Supplier No. HYW0152335G
H-D-Asp(OtBu)-OH is the tert-butyl ester-protected D-aspartic acid with a free N-terminus, supplied as a high-purity solid for use as a peptide synthesis building block and in organic synthesis.
  • Protected D-aspartic acid for solid-phase and solution-phase peptide synthesis.
  • High purity suitable for research and development applications.
  • Tert-butyl side-chain protection provides stability during coupling and deprotection workflows.
  • Available in small lab pack sizes to support method development and scale-up.
  • Well characterized: CAS 64960-75-4; molecular weight 189.21; formula C8H15NO4.