Learn More
Medchemexpress LLC H-D-cis-Hyp-OH (cis-4-hydroxy-D-proline) | 2584-71-6 | 131.13 | 1 G
SDP
Catalog No. 5000446154
Encompass_Preferred
H-D-cis-Hyp-OH (cis-4-hydroxy-D-proline) is a proline derivative involved in bacterial metabolic pathways.
  • Serves as a substrate for cis-4-hydroxy-D-proline dehydrogenase.
  • Acts as a precursor for synthesizing conformationally constrained PNA adenine monomers.
  • Utilized to investigate the specificity and kinetics of D-alanine dehydrogenase.
Catalog No. 50-004-46154 Supplier Medchemexpress LLC Supplier No. HYW0081291G
H-D-cis-Hyp-OH (cis-4-hydroxy-D-proline) is a proline derivative involved in bacterial metabolic pathways.
  • Serves as a substrate for cis-4-hydroxy-D-proline dehydrogenase.
  • Acts as a precursor for synthesizing conformationally constrained PNA adenine monomers.
  • Utilized to investigate the specificity and kinetics of D-alanine dehydrogenase.