Learn More
Medchemexpress LLC H-D-cis-Hyp-OH (cis-4-hydroxy-D-proline) | 2584-71-6 | 131.13 | 100 G
SDP
Catalog No. 5000448748
Encompass_Preferred
H-D-cis-Hyp-OH is a proline derivative involved in bacterial metabolic pathways and serves as a substrate for cis-4-hydroxy-D-proline dehydrogenase.
  • Precursor for synthesizing conformationally constrained PNA adenine monomers.
  • Can be utilized to investigate the specificity and kinetics of D-alanine dehydrogenase.
Catalog No. 50-004-48748 Supplier Medchemexpress LLC Supplier No. HYW008129100G
H-D-cis-Hyp-OH is a proline derivative involved in bacterial metabolic pathways and serves as a substrate for cis-4-hydroxy-D-proline dehydrogenase.
  • Precursor for synthesizing conformationally constrained PNA adenine monomers.
  • Can be utilized to investigate the specificity and kinetics of D-alanine dehydrogenase.