Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eMolecules​ 19898-41-0 | H-D-Glu(Obzl)-Obzl | MFCD00153435 | 5g

Supplier:  eMolecules​ 598443428

Encompass_Preferred

Zinc carbonate | Ambeed | 3486-35-9 | MFCD00041957 | 125.390 | CO3Zn | 57.000 | [Zn++].[O-]C([O-])=O | 100g | 808219646

If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool<|a>

Catalog No. 50-213-3995


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.