Learn More
Medchemexpress LLC H-D-Lys(Z)-OH | 34404-32-5 | MFCD00063120 | 99.6% | 280.32 g/mol | C14H20N2O4 | 25 G
SDP
Catalog No. 5000304090
Encompass_Preferred
H-D-Lys(Z)-OH is the benzyloxycarbonyl (Z/Cbz)-protected D-lysine used as a protected amino acid building block for peptide synthesis and related biochemical research. Supplied as a white to off-white solid with high purity, it is intended for use in coupling reactions and synthetic workflows where side-chain protection is required.
  • Z (Cbz) side-chain protection preserves the lysine ε-amino group during peptide coupling.
  • High purity suitable for peptide synthesis and analytical applications.
  • Solid, white to off-white form for easy handling and accurate weighing.
  • Stable under recommended storage conditions for extended shelf life.
  • Available in multiple laboratory-friendly pack sizes for scalable workflows.
Catalog No. 50-003-04090 Supplier Medchemexpress LLC Supplier No. 25GHYW013850
H-D-Lys(Z)-OH is the benzyloxycarbonyl (Z/Cbz)-protected D-lysine used as a protected amino acid building block for peptide synthesis and related biochemical research. Supplied as a white to off-white solid with high purity, it is intended for use in coupling reactions and synthetic workflows where side-chain protection is required.
  • Z (Cbz) side-chain protection preserves the lysine ε-amino group during peptide coupling.
  • High purity suitable for peptide synthesis and analytical applications.
  • Solid, white to off-white form for easy handling and accurate weighing.
  • Stable under recommended storage conditions for extended shelf life.
  • Available in multiple laboratory-friendly pack sizes for scalable workflows.