Learn More
Bachem H-D-Phe-Cys-Tyr-D-Trp-Arg-Thr-Pen-Thr-NH2
SDP
Supplier:  Bachem 4027138.0005
Encompass_Preferred
H-D-Phe-Cys-Tyr-D-Trp-Arg-Thr-Pen-Thr-NH2, 103429-32-9, C51H69N13O11S2, Mr 1104.32, 5mg. CTAP. CTAP, an analog of CTOP, is a selective u-opioid receptor antagonist.
Catalog No. 50-259-766