Learn More
Bachem H-D-Phe-Cys-Tyr-D-Trp-Orn-Thr-Pen-Thr-NH2
SDP
Supplier:  Bachem 4025623.0005
Encompass_Preferred
H-D-Phe-Cys-Tyr-D-Trp-Orn-Thr-Pen-Thr-NH2 Trifluoroacetate, 103429-31-8, C50H67N11O11S2, Mr 1062.28, 5mg. CTOP, (Tyr3,Orn5,Pen7)-Octreotate amide, NTB (Naltriben). The somatostatin analog CTOP is a very potent and highly selective ligand for u-opioid receptors. CTOP shows only little interaction with other opioid receptor systems.
Catalog No. 50-259-577