Learn More
Bachem H-D-Phe-Phe-Arg-chloromethylketone
SDP
Supplier:  Bachem 4014271.0100
Encompass_Preferred
H-D-Phe-Phe-Arg-chloromethylketone Trifluoroacetate, 74392-49-7, C25H33ClN6O3, Mr 501.03, 100mg. DPhe-Phe-Arg-chloromethylketone. Kallikrein inhibitor which is even effective in preventing apolipoprotein B-100 proteolysis in dextran sulfate (DS)-precipitated human plasma low density lipoproteins (LDL). D-FFR-CMK also inhibits gingipain R.
Catalog No. 50-260-530