Learn More
Medchemexpress LLC HY-123275B 5mg Medchemexpress, H-D-Phe-Pip-Arg-pNA (acetate) CAS:115388-96-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY123275B5MG
Encompass_Preferred
Medchemexpress, HY-123275B 5mg H-D-Phe-Pip-Arg-pNA (acetate) CAS:115388-96-0 H-D-Phe-Pip-Arg-pNA (S-2238) acetate, a chromogenic substrate, is patterned after the N-terminal portion of the A alpha chain of fibrinogen, which is the natural substrate of thrombin. H-D-Phe-Pip-Arg-pNA acetate is specific for thrombin and is used to measure antithrombin-heparin cofactor (AT-III). The AT-III assay using H-D-Phe-Pip-Arg-pNA acetate is sensitive, accurate, and easy to perform. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1010