Learn More
Bachem H-D-Phe-Pro-Arg-chloromethylketone
SDP
Supplier:  Bachem 4033019.0025
Encompass_Preferred
H-D-Phe-Pro-Arg-chloromethylketone Trifluoroacetate, 71142-71-7, C21H31ClN6O3, Mr 450.97, 25mg. PPACK. PPACK, fPR-CMK, a highly effective irreversible inhibitor of thrombin and gingipain R.
Catalog No. 50-260-517