Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-D-Phg(4-Cl)-OH | 43189-37-3 | MFCD03839815 | >98.0% | 185.61 g/mol | C8H8ClNO2 | 10 G
SDP

Catalog No. 5000209575
Encompass_Preferred

H-D-Phg(4-Cl)-OH is a research-grade amino acid derivative (a 4-chloro-phenylglycine derivative) provided as a solid for laboratory peptide synthesis and biochemical research. It is used as a building block in peptide synthesis, small-molecule studies, and other research applications.

  • Amino acid derivative (4-chloro-phenylglycine).
  • Used as a peptide building block and research reagent.
  • CAS: 43189-37-3; formula: C8H8ClNO2; molecular weight: 185.61 g/mol.
  • Physical form: white to off-white solid.
  • Purity reported between 97.0% and >98.0% depending on source and batch.

Catalog No. 50-002-09575 Supplier Medchemexpress LLC Supplier No. HYW04200810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-D-Phg(4-Cl)-OH is a research-grade amino acid derivative (a 4-chloro-phenylglycine derivative) provided as a solid for laboratory peptide synthesis and biochemical research. It is used as a building block in peptide synthesis, small-molecule studies, and other research applications.

  • Amino acid derivative (4-chloro-phenylglycine).
  • Used as a peptide building block and research reagent.
  • CAS: 43189-37-3; formula: C8H8ClNO2; molecular weight: 185.61 g/mol.
  • Physical form: white to off-white solid.
  • Purity reported between 97.0% and >98.0% depending on source and batch.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.