Learn More
Bachem H-D-Pro-Phe-Arg-pNA · 2 HCl
SDP
Catalog No. 50260440
Encompass_Preferred
H-D-Pro-Phe-Arg-pNA · 2 HCl, 62354-56-7, C26H34N8O5 · 2 HCl, Mr 611.53, 25mg. pFR-pNA, chromogenic substrate for the determination of plasma kallikrein-like activity. CAS Number (net): 64816-19-9.
Catalog No. 50-260-440 Supplier Bachem Supplier No. 4029234.0025
Add to Cart
Add to Cart
H-D-Pro-Phe-Arg-pNA · 2 HCl, 62354-56-7, C26H34N8O5 · 2 HCl, Mr 611.53, 25mg. pFR-pNA, chromogenic substrate for the determination of plasma kallikrein-like activity. CAS Number (net): 64816-19-9.