Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-D-Pyr-OH D-Pyroglutamic Aci, Advanced ChemTech

Catalog No. 01338360
Click to view available options
Quantity:
1 g
5 g

Catalog No. 01-338-360

Supplier: Advanced Chem Tech YU32445G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4042-36-8
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
InChI Key ODHCTXKNWHHXJC-UHFFFAOYNA-N
IUPAC Name 5-oxopyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCC(=O)N1

Specifications

Color White/Off-White
Quantity 5 g
Grade Biochemical
Chemical Name or Material H-D-Pyr-OH D-Pyroglutamic Aci
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.