Learn More
Bellco Glass, Inc. H.D. Recprocating Shaker Complete, 115V With 16 Sq Tray& Liner"
SDP
Catalog No. 501945967
Encompass_Preferred
The unit is a laboratory shaker with a reciprocating stroke adjustable for 1”, 1 ¾” & 2 ½”. It is designed to shake clear plastic culture bags with working volumes between 5 & 10 liters, as well as function as a general purpose laboratory shaker. The unit incorporates a digital speed control and 50W brushless DC motor and runs on plastic raceways for minimal noise. Speed is controlled with a numbered dial located on the front of the unit and ranges from 5 to 150 cycles/min approximately. Units are powered by standard 24V in-line power supplies and are CE compliant. The OptimaTM bag trays are sold separately. MADE IN THE USA
Catalog No. 50-194-5967 Supplier Bellco Glass, Inc. Supplier No. 764430115
The unit is a laboratory shaker with a reciprocating stroke adjustable for 1”, 1 ¾” & 2 ½”. It is designed to shake clear plastic culture bags with working volumes between 5 & 10 liters, as well as function as a general purpose laboratory shaker. The unit incorporates a digital speed control and 50W brushless DC motor and runs on plastic raceways for minimal noise. Speed is controlled with a numbered dial located on the front of the unit and ranges from 5 to 150 cycles/min approximately. Units are powered by standard 24V in-line power supplies and are CE compliant. The OptimaTM bag trays are sold separately. MADE IN THE USA