Learn More
Apexbio Technology LLC H-D-Thr-OMe·HCl 60538-15-0 100g
SDP
Catalog No. 5000093160
Encompass_Preferred
H-D-Thr-OMe-HCl is an amino acid derivative containing the methyl ester form of D-threonine characterized by structural stability and defined stereochemical orientation Its molecular characteristics make it suitable for research involving stereoselective synthesis and chiral discrimination This compound serves as a substrate analogue in mechanistic studies of enzyme-catalyzed reactions enabling evaluation of substrate selectivity and enzyme specificity H-D-Thr-OMe-HCl is also applied in studies of structure-function relationships within peptide and protein chemistry aiding investigation of enzyme-substrate interactions and supporting peptide-based therapeutic development
Catalog No. 50-000-93160 Supplier Apexbio Technology LLC Supplier No. A6738100000
H-D-Thr-OMe-HCl is an amino acid derivative containing the methyl ester form of D-threonine characterized by structural stability and defined stereochemical orientation Its molecular characteristics make it suitable for research involving stereoselective synthesis and chiral discrimination This compound serves as a substrate analogue in mechanistic studies of enzyme-catalyzed reactions enabling evaluation of substrate selectivity and enzyme specificity H-D-Thr-OMe-HCl is also applied in studies of structure-function relationships within peptide and protein chemistry aiding investigation of enzyme-substrate interactions and supporting peptide-based therapeutic development