Learn More
Bachem H-D-Val-Leu-Lys-AMC
SDP
Supplier:  Bachem 4008009.0010
Encompass_Preferred
H-D-Val-Leu-Lys-AMC Acetate, 148168-23-4, C27H41N5O5, Mr 515.65, 10mg. D-VLK-AMC, sensitive, highly specific fluorescent substrate for plasmin.
Catalog No. 50-260-309