Learn More
AdipoGen H-D-Val-Leu-Lys-pNA 2HCl
SDP
Supplier:  AdipoGen AGCP30042M005
Encompass_Preferred
Chemical. CAS 62354-43-2. Formula C23H38N6O5 . 2HCl. MW 478.6 . 72.9. D-Val-Leu-Lys-p-nitroanilide is a selective colorimetric substrate for plasmin used to determine the formation of plasmin from plasminogen in amidolytic activity assays and plasminogen activating assays. Plasmin is a plasma serine protease whose main role is to dissolve fibrin blood clots. After cleavage by plasmin the protease activity is quantified by the release of p-nitroaniline pNA from the substrate. The rate of pNA formation, e.g. the increase in absorbance per second at 405 nm, is proportional to the enzymatic activity.
Catalog No. 50-256-2421