Learn More
Medchemexpress LLC H-DL-Cys.HCl | 10318-18-0 | MFCD00064552 | >98.0% | 157.62 | 25 G
SDP
Catalog No. 5000366705
Encompass_Preferred
H-DL-Cys.HCl is a cysteine derivative used for research purposes. Amino acids and amino acid derivatives, such as H-DL-Cys.HCl, are commercially used as ergogenic supplements that influence the secretion of anabolic hormones, supply fuel during exercise, improve mental performance during stress-related tasks, and prevent exercise-induced muscle damage.
  • Used as an ergogenic supplement
  • Influences the secretion of anabolic hormones
  • Supplies fuel during exercise
  • Improves mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
Catalog No. 50-003-66705 Supplier Medchemexpress LLC Supplier No. 25GHYW013651
H-DL-Cys.HCl is a cysteine derivative used for research purposes. Amino acids and amino acid derivatives, such as H-DL-Cys.HCl, are commercially used as ergogenic supplements that influence the secretion of anabolic hormones, supply fuel during exercise, improve mental performance during stress-related tasks, and prevent exercise-induced muscle damage.
  • Used as an ergogenic supplement
  • Influences the secretion of anabolic hormones
  • Supplies fuel during exercise
  • Improves mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage