Learn More
Newcomer Supply Hematoxylin and Eosin (H&E) Control Slides, 98/set
SDP
Catalog No. NC1530514
Encompass
Newcomer Supply Hematoxylin & Eosin (H&E) Control Slides are for the clear demonstration of nuclei, epithelium, connective tissue and muscle, important tissue elements for documentation of high quality nuclear and cytoplasmic staining. Hematoxylin & Eosin (H&E) Control Slides contain sections of positive staining small intestine.
This item is not returnable. View return policy
Catalog No. NC1530514 Supplier Newcomer Supply Supplier No. 4278B
Newcomer Supply Hematoxylin & Eosin (H&E) Control Slides are for the clear demonstration of nuclei, epithelium, connective tissue and muscle, important tissue elements for documentation of high quality nuclear and cytoplasmic staining. Hematoxylin & Eosin (H&E) Control Slides contain sections of positive staining small intestine.