Learn More
Carbosynth LLC. H-Glu(Edans)-Pro-Leu-Phe-Ala-Glu-Arg-Lys(Dabcyl)-OH, 1mg, MW: 1488.74, Molecular formula: C72H97N17O16S
SDP
Supplier:  Carbosynth LLC. SFQ3914PI1MG
Encompass_Preferred
Synonyms: H-Glu(Edans)-Pro-Leu-Phe-Ala-Glu-Arg-Lys(Dabcyl)-OH, Application: Substrate for Calpain 1, Salt Form:, Acetate Form, Storage Temperature: -20°C, References: D. Cuerrier, T. Moldoveanu, P.L. Davies. J. Biol. Chem., 280, 40632 (2005).
Catalog No. 50-168-7086