Learn More
Cayman Chemical h-e5NT InhIbItr 5C 1mg
SDP
Catalog No. 502943112
Encompass_Preferred
An inhibitor of ecto-5'-nucleotidase (IC50s = 0.37 and 1.66 µM for the human and rat enzymes, respectively); inhibits ENPP1, ENPP3, and TNAP (IC50s = 1.12, 1.37, and 1.48 µM, respectively, for the human enzymes)
Catalog No. 50-294-3112 Supplier Cayman Chemical Supplier No. 402661 MG
An inhibitor of ecto-5'-nucleotidase (IC50s = 0.37 and 1.66 µM for the human and rat enzymes, respectively); inhibits ENPP1, ENPP3, and TNAP (IC50s = 1.12, 1.37, and 1.48 µM, respectively, for the human enzymes)