Learn More
Cayman Chemical h-e5NT InhIbItr 5C 5mg
SDP
Catalog No. 502942525
Encompass_Preferred
An inhibitor of ecto-5'-nucleotidase (IC50s = 0.37 and 1.66 µM for the human and rat enzymes, respectively); inhibits ENPP1, ENPP3, and TNAP (IC50s = 1.12, 1.37, and 1.48 µM, respectively, for the human enzymes)
Catalog No. 50-294-2525 Supplier Cayman Chemical Supplier No. 402665 MG
Add to Cart
Add to Cart
An inhibitor of ecto-5'-nucleotidase (IC50s = 0.37 and 1.66 µM for the human and rat enzymes, respectively); inhibits ENPP1, ENPP3, and TNAP (IC50s = 1.12, 1.37, and 1.48 µM, respectively, for the human enzymes)