Learn More
Elemental Scientific Inc H-F RESISTANT MICROFLOW
SDP
Catalog No. NC0692146
Encompass_Preferred
400 uL/min self-aspirating sample capillary for ST nebulizer. (0.5 mm ID, orange marker)
Catalog No. NC0692146 Supplier Elemental Scientific Inc Supplier No. ES2041
400 uL/min self-aspirating sample capillary for ST nebulizer. (0.5 mm ID, orange marker)