Learn More
Genscript Corporation h-FABP 28D1E mAb Mouse
Catalog No. 5000057337
Encompass
Heart-type fatty acid binding protein (H-FABP) is encoded by the FABP3 gene with a low molecular weight of 15 kDa It is released in large quantities into blood at an early stage of acute myocardial infarction (AMI) H-FABP is a serum marker with high sensitivity and specificity for diagnosing AMI and myocardial ischemia
Catalog No. 50-000-57337 Supplier Genscript Corporation Supplier No. V009021
Heart-type fatty acid binding protein (H-FABP) is encoded by the FABP3 gene with a low molecular weight of 15 kDa It is released in large quantities into blood at an early stage of acute myocardial infarction (AMI) H-FABP is a serum marker with high sensitivity and specificity for diagnosing AMI and myocardial ischemia