Learn More
Thermo Scientific™ H-Flex HS4 Swinging Bucket Rotor with SlideCoat for 4 L General Purpose X Pro and ST Plus Centrifuges
Catalog No. 75003330 Shop All Thermo Scientific Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
75-003-330 Autoclavable
1 options
Catalog No. 75-003-330 Supplier Thermo Scientific™ Supplier No. 75003330
  • $6,490.00
    $4,604.65 / Each of 1
    Save $1,885.35
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote
Spin more with less! High performance and flexibility, all-in-one H-Flex HS4 rotor.
Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor at 7197xg in 4 L High Performance General Purpose Centrifuges .
    • Auto-Lock enable fast, one-handed rotor exchanges
    • Spins blood tubes, conical tubes and microplates without changing rotor or buckets
    • Get results faster without the need to change hardware
    • Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
TRUSTED_SUSTAINABILITY
Specifications
Material Anodized Aluminum
Type Swinging Bucket Rotor
Capacity 16 x 50 mL or 36 x 15 mL, 66 x 10 mL or 84 x 5/7 mL Blood Tubes
Max. RCF 7,197 x g
Max. Speed 6,100 rpm
For Use With (Equipment) Sorvall X4 Pro, Sorvall X4 F Pro, Multifuge X4 Pro, Multifuge X4 F Pro, Sorvall ST4 Plus, Sorvall ST4F Plus, Megafuge ST4 Plus, Megafuge ST4F Plus
Includes Windshield, rotor cross and buckets, 10 microwell microplates, 2 deepwell plates
Biocontainment Yes
Autoclavable Autoclavable