Learn More
Medchemexpress LLC H-Glu(AMC)-OH | 98516-76-8 | MFCD00038085 | 95.0% | 304.30 g/mol | C15H16N2O5 | 25 MG
SDP
Catalog No. 5000281434
Encompass_Preferred
H-Glu(AMC)-OH is an amino acid derivative used as a fluorogenic peptide substrate to measure aminopeptidase A activity. It releases the fluorescent leaving group 7-amino-4-methylcoumarin (AMC) upon enzymatic cleavage, enabling sensitive fluorometric assays for biochemical research. The material is supplied as a powder for laboratory use.
  • Fluorogenic substrate for aminopeptidase A activity assays.
  • Releases 7-amino-4-methylcoumarin (AMC) upon enzymatic cleavage for sensitive fluorescence detection.
  • Powder form suitable for preparation of assay solutions.
  • Stable when stored as a powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • In-solution stability: short-term storage at -80°C (months) or -20°C (weeks) depending on solvent.
  • Molecular formula C15H16N2O5; molecular weight 304.30 g/mol.
Catalog No. 50-002-81434 Supplier Medchemexpress LLC Supplier No. HYW25072525MG
H-Glu(AMC)-OH is an amino acid derivative used as a fluorogenic peptide substrate to measure aminopeptidase A activity. It releases the fluorescent leaving group 7-amino-4-methylcoumarin (AMC) upon enzymatic cleavage, enabling sensitive fluorometric assays for biochemical research. The material is supplied as a powder for laboratory use.
  • Fluorogenic substrate for aminopeptidase A activity assays.
  • Releases 7-amino-4-methylcoumarin (AMC) upon enzymatic cleavage for sensitive fluorescence detection.
  • Powder form suitable for preparation of assay solutions.
  • Stable when stored as a powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • In-solution stability: short-term storage at -80°C (months) or -20°C (weeks) depending on solvent.
  • Molecular formula C15H16N2O5; molecular weight 304.30 g/mol.