Learn More
Medchemexpress LLC H-Glu(AMC)-OH | 98516-76-8 | MFCD00038085 | 98.0% | 304.30 g/mol | C15H16N2O5 | 50 MG
SDP
Catalog No. 5000281633
Encompass_Preferred
H-Glu(AMC)-OH is a fluorogenic amino acid derivative used as a peptide substrate to measure aminopeptidase A enzyme activity. Enzymatic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal that enables quantitative fluorescence-based activity assays. Intended for research use only.
  • Fluorogenic substrate for aminopeptidase A activity assays.
  • Releases 7-amino-4-methylcoumarin (AMC) upon enzymatic cleavage.
  • Enables quantitative fluorescence-based enzyme activity measurements.
  • Available as a solid powder suitable for storage and assay preparation.
  • Molecular weight 304.30 g/mol; formula C15H16N2O5.
  • For research use only; not validated for medical applications.
Catalog No. 50-002-81633 Supplier Medchemexpress LLC Supplier No. HYW25072550MG
H-Glu(AMC)-OH is a fluorogenic amino acid derivative used as a peptide substrate to measure aminopeptidase A enzyme activity. Enzymatic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal that enables quantitative fluorescence-based activity assays. Intended for research use only.
  • Fluorogenic substrate for aminopeptidase A activity assays.
  • Releases 7-amino-4-methylcoumarin (AMC) upon enzymatic cleavage.
  • Enables quantitative fluorescence-based enzyme activity measurements.
  • Available as a solid powder suitable for storage and assay preparation.
  • Molecular weight 304.30 g/mol; formula C15H16N2O5.
  • For research use only; not validated for medical applications.