Learn More
Bachem H-Glu-Glu-OH
SDP
Supplier:  Bachem 4000466.1000
Encompass_Preferred
H-Glu-Glu-OH, 3929-61-1, C10H16N2O7, Mr 276.25, 1g. Dipeptides as aspartylaspartate and glutamylglutamate can be utilized as growth substrates for the oral bacteria Porphyromonas gingivalis, Prevotella intermedia, Prevotella nigrescens and Fusobacterium nucleatum.
Catalog No. NC1775986