Learn More
Bachem H-Glu(H-Lys-OH)-OH
SDP
Supplier:  Bachem 4001034.0250
Encompass_Preferred
H-Glu(H-Lys-OH)-OH, 17105-15-6, C11H21N3O5, Mr 275.31, 250mg. H-Lys(retro-HO-Glu-H)-OH, H-g-Glu-e-Lys-OH. The Glu-Lys isopeptide bond is formed as a crosslink in proteins in normal, not virus-transformed cells. Additionally, the acceleration of fibrin e-(g-glutamyl)lysine (GGEL) crosslinking by red blood cells has been observed. GGEL detected in the CSF is a biomarker of neurodegenerative diseases.
Catalog No. 50-382-080