Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-γ-Glu-Leu-OH | 2566-39-4 | 99.2% | 260.29 | 50 MG
SDP

Catalog No. 5000397533
Encompass_Preferred

H-γ-Glu-Leu-OH (γ-Glu-Leu) is a dipeptide composed of γ-glutamic acid and leucine, featuring a hydroxyl group termination. It is classified as an amino acid derivative and is intended for research use only.

  • Appearance: Solid
  • Color: White to off-white
  • Sequence: γ-Glu-Leu
  • Sequence shortening: γ-EL
  • In vitro solubility: Soluble in DMSO at concentrations of ≥ 100 mg/mL (384.19 mM).
  • Shipping: Room temperature in continental US.
  • Powder storage: Sealed, away from moisture and light, at -80°C for 2 years or -20°C for 1 year.
  • In solvent storage: Sealed, away from moisture and light, at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-003-97533 Supplier Medchemexpress LLC Supplier No. HYP463250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-γ-Glu-Leu-OH (γ-Glu-Leu) is a dipeptide composed of γ-glutamic acid and leucine, featuring a hydroxyl group termination. It is classified as an amino acid derivative and is intended for research use only.

  • Appearance: Solid
  • Color: White to off-white
  • Sequence: γ-Glu-Leu
  • Sequence shortening: γ-EL
  • In vitro solubility: Soluble in DMSO at concentrations of ≥ 100 mg/mL (384.19 mM).
  • Shipping: Room temperature in continental US.
  • Powder storage: Sealed, away from moisture and light, at -80°C for 2 years or -20°C for 1 year.
  • In solvent storage: Sealed, away from moisture and light, at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.